EMAIL THIS PAGE TO A FRIEND
Expert opinion on drug discovery
The influence of lipophilicity in drug discovery and design.
PMID 22992175
Abstract
The role of lipophilicity in drug discovery and design is a critical one. Lipophilicity is a key physicochemical property that plays a crucial role in determining ADMET (absorption, distribution, metabolism, excretion, and toxicity) properties and the overall suitability of drug candidates. There is increasing evidence to suggest that control of physicochemical properties such as lipophilicity, within a defined optimal range, can improve compound quality and the likelihood of therapeutic success. This review focuses on understanding lipophilicity, techniques used to measure lipophilicity, and summarizes the importance of lipophilicity in drug discovery and development, including a discussion of its impact on individual ADMET parameters as well as its overall influence on the drug discovery and design process, specifically within the past 15 years. A current review of the literature reveals a continued reliance on the synthesis of novel structures with increased potency, rather than a focus on maintaining optimal physicochemical properties associated with ADMET throughout drug optimization. Particular attention to the optimum region of lipophilicity, as well as monitoring of lipophilic efficiency indices, may contribute significantly to the overall quality of candidate drugs at different stages of discovery.
Related Materials