EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A near-infrared fluorescent turn-on probe for fluorescence imaging of hydrogen sulfide in living cells based on thiolysis of dinitrophenyl ether.
PMID 22992474
Abstract
We have constructed a novel NIR fluorescent turn-on hydrogen sulfide probe suitable for fluorescent imaging in living cells based on thiolysis of dinitrophenyl ether.