EMAIL THIS PAGE TO A FRIEND
Le infezioni in medicina : rivista periodica di eziologia, epidemiologia, diagnostica, clinica e terapia delle patologie infettive
Necrotizing pneumonia caused by Panton-Valentine leukocidin-producing methicillin-susceptible Staphylococcus aureus (MSSA).
PMID 22992562
Abstract
Staphylococcus aureus harboured by Panton-Valentine leukocidin (PVL) is emerging as a serious problem worldwide. There has been an increase in the incidence of necrotizing lung infections in otherwise healthy young people with very high mortality rate associated with these strains. This report documents a confirmed case of necrotizing pneumonia due to methicillin-susceptible S. aureus (MSSA) harbouring Panton-Valentine leukocidin genes. An apparently healthy 49-year old man was admitted to our hospital for dyspnea and he quickly developed acute respiratory distress syndrome. MSSA harbouring Panton-Valentine leukocidin genes were cultured from the abscess fluid and from multiple blood specimens. Aggressive antibiotic therapy was started and intensive supportive care led finally to a complete recovery. Rapid identification of Panton-Valentine leukocidin in MSSA samples should be supposed when a young, immunocompetent patient, develops a necrotizing pneumonia. Bactericidal antistaphylococcal antibiotics are recommended for the treatment as soon as possible to avoid the potentially devastating consequences of this kind of S. aureus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51454
Methicillin sodium salt, ≥95% (HPLC)
C17H19N2NaO6S