EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Simultaneous quantitative analysis of four lignanoids in Schisandra chinensis by quantitative analysis of multi-components by single marker].
PMID 22993860
Abstract
The aim of the study is to establish a new method of quality evaluation and validate its feasibilities by the simultaneous quantitative assay of four lignanoids in Schisandra chinensis. A new quality evaluation method, quantitative analysis of multi-components by single marker (QAMS), was established and validated with Schisandra chinensis. Four main lignanoids, schisandrin, schisantherin A, deoxyschizandrin and gamma-schizandrin, were selected as analytes and schisandrin as internal reference substance to evaluate the quality. Their contents in 13 different batches of samples, collected from different bathes, were determined by both external standard method and QAMS. The method was evaluated by comparison of the quantitative results between external standard method and QAMS. No significant differences were found in the quantitative results of four lignanoids in 13 batches of S. chinensis determined by external standard method and QAMS. QAMS is feasible for determination of four lignanoids simultaneously when some authentic standard substances were unavailable, and the developed method can be used for quality control of S. chinensis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0054
Schizandrin, ≥98% (HPLC)
C24H32O7