EMAIL THIS PAGE TO A FRIEND
Lakartidningen
[Remove Tavegyl].
PMID 22993901
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0445
Clemastine fumarate salt, ≥98% (HPLC)
C21H26ClNO · C4H4O4