EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
Acute and chronic antiparkinsonian effects of the novel nociceptin/orphanin FQ receptor antagonist NiK-21273 in comparison with SB-612111.
PMID 22994368
Abstract
Nociceptin/orphanin FQ (N/OFQ) peptide (NOP) receptor antagonists have been proposed as a novel therapeutic approach to Parkinson's disease. Main limitations of previous studies were the use of structurally similar compounds and the evaluation of their acute effects only. We report here on the acute and long-term antiparkinsonian effects of the novel compound 2-[3-[4-(2-chloro-6-fluoro-phenyl)-piperidin-1-ylmethyl]-2-(morpholine-4-carbonyl)-indol-1-yl]-acetamide (NiK-21273) in comparison with the potent and selective NOP receptor antagonist SB-612111. Basic pharmacological properties of NiK-21273 were studied in cell lines and isolated tissues (mouse and rat vas deferens). Antiparkinsonian effects were studied in reserpinized mice and 6-hydroxydopamine hemilesioned rats under both acute and chronic administration protocols. In vitro, NiK-21273 behaved as a potent (pA(2) 7.7) and selective NOP receptor antagonist. In vivo, it reduced hypokinesia in reserpinized mice at 0.1 and 1 but not 10 mg·kg(-1), whereas SB-612111 (0.01-1 mg·kg(-1)) provided a dose-dependent antiparkinsonian effect. NiK-21273 ameliorated motor performance in 6-hydroxydopamine hemilesioned rats at 0.5 and 5 but not 15 mg·kg(-1). SB-612111 replicated these effects in the 0.01-1 mg·kg(-1) range without loss of efficacy. Both antagonists synergized with L-DOPA at subthreshold doses. Chronic administration of NiK-21273 provided delayed improvement in baseline activity at 0.5 and 1.5 mg·kg(-1), although tolerance to the higher dose was observed. Conversely, SB-612111 (1 mg·kg(-1)) maintained its effects over time without modifying baseline activity. NOP receptor antagonists provide motor benefit in parkinsonism models although the 'therapeutic' window and long-term effects may vary between compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
83580
Reserpine, crystallized, ≥99.0% (HPLC)
C33H40N2O9