EMAIL THIS PAGE TO A FRIEND
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology
Vaginal atrophy in breast cancer survivors: role of vaginal estrogen therapy.
PMID 22994445
Abstract
Early menopause and related vaginal atrophy is a well known side-effect of hormone adjuvant treatment in breast cancer patients, particularly during aromatase-inhibitors therapy. Due to estrogens contra-indication, proper therapy for such symptom remains often an inadequately addressed clinical problem. After an accurate assessment of the risk/benefit ratio, vaginal low-dose estrogen treatment (better with estriol) [corrected] may have a role in controlling vaginal atrophy in selected and informed breast cancer women.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E3750
17-Epiestriol
C18H24O3