EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Biogenic phosphorus compounds in sediment and suspended particles in a shallow eutrophic lake: a ³¹P-nuclear magnetic resonance (³¹P NMR) study.
PMID 22994917
Abstract
Differences in biogenic phosphorus (P) compounds between sediment and suspended particles in aquatic environments are important for understanding the mechanisms of internal P loading, but these differences are still unknown. We used solution-state (31)P-nuclear magnetic resonance spectroscopy ((31)P NMR) with NaOH-ethylenediaminetetraacetic extraction to detect the multiple P compounds in suspended particles and sediment in the eutrophic Lake Kasumigaura, including orthophosphate monoesters, orthophosphate diesters, pyrophosphate, and polyphosphate. We tested the hypothesis that there is a significant difference between these groups in suspended particles and sediment. Biogenic P other than orthophosphate was found in significantly higher proportions in suspended particles (74.3% of total P) than in sediment (25.6%). Orthophosphate monoesters were comparatively more abundant in suspended particles, as indicated by the ratio of orthophosphate diesters to monoesters (average, 0.31 for suspended particles; 1.05 for sediment). The compounds identified as orthophosphate monoesters by (31)P NMR spectroscopy originated mainly from phospholipids (α-glycerophosphate and β-glycerophosphate) and ribonucleic acid (RNA-P), whereas the orthophosphate diesters included mostly DNA (DNA-P). These results suggest that the dynamics of orthophosphate diesters, the production of DNA-P, or the degradation of phospholipids, play an important role in P cycling in Lake Kasumigaura.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17766
DL-α-Glycerol phosphate magnesium salt hydrate, ~85% (KT)
C3H7MgO6P · xH2O