EMAIL THIS PAGE TO A FRIEND
International journal of occupational safety and ergonomics : JOSE
New filtering antimicrobial nonwovens with various carriers for biocides as respiratory protective materials against bioaerosol.
PMID 22995135
Abstract
This study evaluated the bioactivity of polypropylene melt-blown filtering nonwovens used in respiratory protective devices (RPD) with a biocidal agent (alkylammonium microbiocides) on 2 mineral carriers. Two types of carriers were tested: a bentonite, with an aluminosilicate base, and a perlite, volcanic glass. High biostatic and biocidal effects of modified nonwovens with biocides were tested against Escherichia coli (E. coli) and Staphylococcus aureus (S. aureus) bacteria. Nonwovens modified with a biocide on a bentonite carrier showed an opposite reaction to a biocide on a perlite. The research also showed that 10% concentration of a biocidal agent on a perlite carrier was sufficient to inhibit the growth of bacteria (100% reduction) placed in the structure of a filtering material during normal use of RPD. A comparison of the biological activity of 2 filtering materials, each containing 10% of a perlite and produced in a laboratory and industrial conditions, showed no statistically significant differences.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
285234 Bentonite
682659 Nanoclay, hydrophilic bentonite