EMAIL THIS PAGE TO A FRIEND
Obstetrics and gynecology
Committee opinion no. 539: adolescents and long-acting reversible contraception: implants and intrauterine devices.
PMID 22996129
Abstract
Long-acting reversible contraception (LARC)—intrauterine devices and the contraceptive implant—are safe and appropriate contraceptive methods for most women and adolescents. The LARC methods are top-tier contraceptives based on effectiveness, with pregnancy rates of less than 1% per year for perfect use and typical use. These contraceptives have the highest rates of satisfaction and continuation of all reversible contraceptives. Adolescents are at high risk of unintended pregnancy and may benefit from increased access to LARC methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32809
Desogestrel, VETRANAL, analytical standard
C22H30O
SML0356
Etonogestrel, ≥98% (HPLC)
C22H28O2