EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthesis and biological activity of an acyclic analogue of 5,6,7,8-tetrahydrofolic acid, N-[4-[[3-(2,4-diamino-1,6-dihydro-6-oxo-5- pyrimidinyl)propyl]amino]-benzoyl]-L-glutamic acid.
PMID 2299624
Abstract
The synthesis and biological evaluation of N-[4-[[3-(2,4-diamino-1,6-dihydro-6-oxo-5-pyrimidinyl)propyl]amino]- benzoyl]-L-glutamic acid (1) (5-DACTHF, 543U76), an acyclic analogue of 5,6,7,8-tetrahydrofolic acid (THFA), are described. The key intermediate, hemiaminal 8, was prepared in four stages from 3-chloropropionaldehyde diethyl acetal. Reaction of 8 with dimethyl N-(4-aminobenzoyl)-L-glutamate gave the 2,4-bis(acetylamino) derivative 11, which was hydrolyzed with 1 N sodium hydroxide to give 1; the glycine analogue 16 was prepared in a similar manner. The N-methyl analogue 2 and N-formyl analogue 3 were prepared from 11 and 1, respectively. Compounds 1-3 inhibited growth of Detroit 98 and L cells in cell culture, with IC50s ranging from 2 to 0.018 microM. Cell culture toxicity reversal studies and enzyme inhibition tests showed that 1 was cytotoxic but not by the mechanism of the dihydrofolate reductase inhibitor aminopterin. Compound 1 and its polyglutamylated homologues inhibited glycinamide ribonucleotide transformylase (GAR-TFase) and aminoimidazole ribonucleotide transformylase (AICAR-TFase), the folate-dependent enzymes in de novo purine biosynthesis; and 1 was an effective substrate for mammalian folyl-polyglutamate synthetase. The compound inhibited (IC50 = 20 nM) the conversion of [14C]formate to [14C]-formylglycinamide ribonucleotide by MOLT-4 cells in culture. These data suggest that the site of action of 1 is inhibition of purine de novo biosynthesis. Moderate activity was observed against P388 leukemia in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
26130
3-Chloropropionaldehyde diethylacetal, technical, ≥90% (GC)
C7H15ClO2