EMAIL THIS PAGE TO A FRIEND
TheScientificWorldJournal
NOD2 agonist promotes the production of inflammatory cytokines in VSMC in synergy with TLR2 and TLR4 agonists.
PMID 22997500
Abstract
To investigate the expression of NOD2 in human VSMCs, its role in the production of inflammatory cytokines in VSMC and the possible interaction of NOD2-mediated signaling pathway with those mediated by TLR2 and TLR4. Human coronary artery smooth muscle cells were stimulated with NOD2 agonist MDP alone or in combination with either TLR2 agonist PAM3 or TLR4 agonist LPSs. The mRNA expression of NOD2 and FGF-2 were measured by RT-PCR. The concentration of IL-8 and TNF-α in the culture supernatants was determined by ELISA. VSMC proliferation ability was analyzed by MTT assay. MDP up regulated the expression of NOD2 mRNA in VSMC in a time-dependent manner, up regulated the expression of FGF-2 mRNA in VSMC, induced the production of IL-8 and TNF-α, and promoted the proliferation of VSMC. Additionally, MDP synergied with LPS and PAM3 to promote the proliferation of VSMC and induce the production of IL-8 and TNF-α. The activation of NOD2-mediated innate immune signaling pathway can increase the proliferation ability of VSMC and induce the production of inflammatory cytokines in VSMC. It is also shown a synergistic effect with TLR2- and TLR4-mediated signaling pathways in this process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9519
N-Acetylmuramyl-L-alanyl-D-isoglutamine hydrate, ≥98% (TLC)
C19H32N4O11 · xH2O