EMAIL THIS PAGE TO A FRIEND
The Journal of chemical physics
High resolution 11B NMR of magnesium diboride using cryogenic magic angle spinning.
PMID 22998255
Abstract
Static and magic-angle spinning (11)B nuclear magnetic resonance (NMR) data at 4.7 T and 8.5 T have been obtained under cryogenic conditions on a diluted sample of magnesium diboride powder in the normal and superconducting state. The data provide accurate information on the magnetic shift and longitudinal relaxation time down to a temperature of 8 K, with a resolution improvement over the entire temperature range. The onset of superconductivity is unaffected by the sample rotation, as revealed by a steep variation of the magnetic shift just below the critical temperature.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
553913
Magnesium boride, powder, −100 mesh, ≥99% trace metals basis
B2Mg