EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Trifluoromethylation of α-haloketones.
PMID 22998369
Abstract
The C-X bond (X = Br, Cl) of α-haloketones is smoothly trifluoromethylated with the fluoroform-derived CuCF(3) reagent recently developed in our laboratories. This is the first nucleophilic α-trifluoromethylation reaction of carbonyl compounds and a rare example of CF(3)-C(sp(3)) coupling. The transformation employs only low-cost chemicals and cleanly occurs in up to 99% yield at room temperature, thereby providing an unprecedentedly easy entry to valuable 2,2,2-trifluoroethylketones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295337
Trifluoromethane, ≥98%
CHF3