EMAIL THIS PAGE TO A FRIEND
Clinical biochemistry
Purine metabolites in fibromyalgia syndrome.
PMID 23000315
Abstract
To evaluate serum purine metabolite concentrations in patients affected by fibromyalgia syndrome (FMS) and the relationships between their levels and FM clinical parameters. Serum purine levels were quantified using LC/UV-vis in 22 fibromyalgic females (according to the American College of Rheumatology classification criteria) and 22 healthy females. Significantly higher serum inosine, hypoxanthine and xanthine levels (p<0.001) and significantly lower serum adenosine (p<0.05) were detected in the FMS patients vs healthy controls. Our data show a negative correlation between adenosine and Fibromyalgia Impact Questionnaire (FIQ). Study results suggest that purines, in particular adenosine and inosine, may be involved in pain transmission in fibromyalgia.