EMAIL THIS PAGE TO A FRIEND
The international journal of biochemistry & cell biology
Phosphatidylcholine-specific phospholipase C/heat shock protein 70 (Hsp70)/transcription factor B-cell translocation gene 2 signaling in rat bone marrow stromal cell differentiation to cholinergic neuron-like cells.
PMID 23000394
Abstract
Although bone marrow stromal cells (BMSCs) can differentiate into neuron-like cells, the mechanisms underlying neuronal differentiation are not well understood. We recently found that inhibition of phosphatidylcholine-specific phospholipase C (PC-PLC) by its inhibitor D609 promoted BMSCs' differentiation into cholinergic neuron-like cells. Using the effective small molecule D609 and gene microarray technology, we investigated the change of gene expression profile to identify key mediators involved in the neuronal differentiation. We selected heat shock protein 70 (Hsp70) and transcription factor B-cell translocation gene 2 (Btg2) that were maximally up-regulated for further study. We found that functional suppression of Hsp70 blocked D609-induced increase of Btg2 expression and cholinergic neuronal differentiation of BMSCs. These results demonstrated that Hsp70 was the pivotal factor in PC-PLC-medicated neuronal differentiation of BMSCs, and Btg2 might be its downstream target. Our findings provide new clues for controlling BMSCs' differentiation into cholinergic neuron-like cells and provide a putative strategy for neurodegenerative diseases therapies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8543
O-Tricyclo[5.2.1.02,6]dec-9-yl dithiocarbonate potassium salt, ≥95%, solid
C11H15KOS2