EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Separation of fluorescently labeled phosphoinositides and sphingolipids by capillary electrophoresis.
PMID 23000742
Abstract
Phosphoinositides (PIs) and sphingolipids regulate many aspects of cell behavior and are often involved in disease processes such as oncogenesis. Capillary electrophoresis with laser induced fluorescence detection (CE-LIF) is emerging as an important tool for enzymatic assays of the metabolism of these lipids, particularly in cell-based formats. Previous separations of phosphoinositide lipids by CE required a complex buffer with polymer additives which had the disadvantages of high cost and/or short shelf life. Further a simultaneous separation of these classes of lipids has not been demonstrated in a robust buffer system. In the current work, a simple separation buffer based on NaH(2)PO(4) and 1-propanol was optimized to separate two sphingolipids and multiple phosphoinositides by CE. The NaH(2)PO(4) concentration, pH, 1-propanol fraction, and a surfactant additive to the buffer were individually optimized to achieve simultaneous separation of the sphingolipids and phosphoinositides. Fluorescein-labeled sphingosine (SFL) and sphingosine 1-phosphate (S1PFL), fluorescein-labeled phosphatidyl-inositol 4,5-bisphosphate (PIP2) and phosphatidyl-inositol 3,4,5-trisphosphate (PIP3), and bodipy-fluorescein (BFL)-labeled PIP2 and PIP3 were separated pairwise and in combination to demonstrate the generalizability of the method. Theoretical plate numbers achieved were as high as 2×10(5) in separating fluorophore-labeled PIP2 and PIP3. Detection limits for the 6 analytes were in the range of 10(-18)-10(-20)mol. The method also showed high reproducibility, as the relative standard deviation of the normalized migration time for each analyte in the simultaneous separation of all 6 compounds was less than 1%. The separation of a mixture composed of diacylglycerol (DAG) and multiple phosphoinositides was also demonstrated. As a final test, fluorescent lipid metabolites formed within cells loaded with BFLPIP2 were separated from a cell lysate as well as a single cell. This simple and robust separation method for SFL and S1PFL and various metabolites of phosphoinositide-related signal transduction is expected to enable improved enzymatic assays for biological and clinical applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34871
1-Propanol, for HPLC, ≥99.9%
C3H8O
402893
1-Propanol, ACS reagent, ≥99.5%
C3H8O
33538
1-Propanol, puriss. p.a., Reag. Ph. Eur., ≥99.5% (GC)
C3H8O
24135
1-Propanol, ≥99% (GC), purum
C3H8O
S9763
Sodium phosphate dibasic, ACS reagent, ≥99.0%
HNa2O4P
71640
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
HNa2O4P
04276
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
HNa2O4P
71650
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
HNa2O4P · 12H2O
04273
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-101% (calc. to the dried substance)
HNa2O4P · 12H2O
30414
Sodium phosphate dibasic dodecahydrate, 99-101% (calc. to the dried substance)
HNa2O4P · 12H2O
431478
Sodium phosphate dibasic heptahydrate, ≥99.99% trace metals basis
HNa2O4P · 7H2O
S9390
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
HNa2O4P · 7H2O
S2429
Sodium phosphate dibasic heptahydrate, meets USP testing specifications
HNa2O4P · 7H2O
71911
Sodium phosphate tribasic dodecahydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Na3O4P · 12H2O
222003
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Na3O4P · 12H2O