EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Polymorphism and tautomeric preference in fenobam and the utility of NLO response to detect polymorphic impurities.
PMID 23000909
Abstract
Crystal structures of polymorphs and solvatomorphs of the potential anxiolytic drug fenobam exhibit an exclusive preference for one of the two possible tautomeric structures. A novel methodology based on nonlinear optical response has been successfully employed to detect the presence of a polymorphic impurity in a mixture of polymorphs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0430
Fenobam, ≥98% (HPLC), solid
C11H11ClN4O2