EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Conformational modulation of Ant-Pro oligomers using chirality alteration of proline residues.
PMID 23001178
Abstract
Structural modulation of Ant-Pro (anthranilic acid-proline) oligomers has been carried out by chirality alteration of the proline residues. The results suggest that the chirality altered oligomers show well-defined helical conformation featuring nine-membered hydrogen bonding interactions - without compromising conformational rigidity.