EMAIL THIS PAGE TO A FRIEND
Cellular & molecular biology letters
Evaluation of melanogenesis in A-375 melanoma cells treated with 5,7-dimethoxycoumarin and valproic acid.
PMID 23001511
Abstract
Malignant melanoma (melanoma malignum) is one of the most dangerous types of tumor. It is very difficult to cure. In recent years, a lot of attention has been given to chemoprevention. This method uses natural and synthetic compounds to interfere with and inhibit the process of carcinogenesis. In this study, a new treatment strategy was proposed consisting of a combination of 5,7-dimethoxycoumarin (DMC), an activator of melanogenesis, and valproic acid (VPA), a well-known drug that is one of the histone deacetylase inhibitors (HDACis). In conjunction with 1 mM VPA, all of the tested concentrations of DMC (10-150 μM) significantly decreased the proliferation of A-375 cells. VPA and DMC also induced the synthesis of melanin and the formation of dendrite and star-shaped cells. Tyrosinase gene expression and tyrosinase activity significantly increased in response to VPA treatment. Pyrolysis with gas chromatography and mass spectrometry (Py-GC/MS) was used to investigate the structure of the isolated melanin. This showed that the quantitative and qualitative components of melanin degradation products are dependent on the type of applied melanogenesis inductor. Products derived from eumelanin were detected in the pyrolytic profile of melanin isolated from A-375 cells stimulated with DMC. Thermal degradation of melanin isolated from melanoma cells after exposure to VPA or a mixture of VPA and DMC revealed the additional presence of products derived from pheomelanin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
116238
5,7-Dimethoxycoumarin, 98%
C11H10O4