EMAIL THIS PAGE TO A FRIEND
Physical review letters
Gelation via ion exchange in discotic suspensions.
PMID 23004332
Abstract
The phase behavior of charged disk suspensions displays a strong dependence on ionic strengths, as the interplay between excluded volume and electrostatic interactions determines the formation of glasses, gels, and liquid crystal states. The various ions in natural soil or brine, however, could present additional effects, especially considering that most platelet structures bear a momentous ion-exchange capacity. Here we observed how ion exchange modulates and controls the interaction between individual disks and leads to unconventional phase transitions from isotropic gel to nematic gel and finally to nematic liquid crystals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
651664
Tetrabutylammonium azide
C16H36N4
86853
Tetrabutylammonium bisulfate, for ion pair chromatography, ≥99.0%
C16H37NO4S
86868
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
C16H37NO4S
72453
Tetrabutylammonium bisulfate solution, ~55% in H2O
C16H37NO4S
98479
Tetrabutylammonium bisulfate solution, for ion pair chromatography, concentrate, ampule
C16H37NO4S
86857
Tetrabutylammonium bromide, for ion pair chromatography, ≥99.0%
C16H36BrN
193119
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
C16H36BrN
462144
Tetrabutylammonium bromide solution, 50 wt. % in H2O
C16H36BrN
86852
Tetrabutylammonium chloride, for ion pair chromatography, ≥99.0%
C16H36ClN
86870
Tetrabutylammonium chloride, ≥97.0% (NT)
C16H36ClN
86862
Tetrabutylammonium chloride solution, for ion pair chromatography, concentrate, ampule
C16H36ClN
358665
Tetrabutylammonium cyanide, 95%
C17H36N2
79989
Tetrabutylammonium cyanide, technical, ≥80%
C17H36N2
216143
Tetrabutylammonium fluoride solution, 1.0 M in THF
C16H36FN
361399
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
C16H36FN
155837
Tetrabutylammonium hydrogensulfate, 97%
C16H37NO4S
86854
Tetrabutylammonium hydroxide solution, ~40% in water, for ion chromatography
C16H37NO
86863
Tetrabutylammonium hydroxide solution, 54.0-56.0% in H2O
C16H37NO
86880
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
C16H37NO
178780
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
C16H37NO
86903
Tetrabutylammonium iodide, for ion pair chromatography, ≥99.0%
C16H36IN
86912
Tetrabutylammonium iodide, for electrochemical analysis, ≥99.0%
C16H36IN
86890
Tetrabutylammonium iodide, ≥99.0% (AT)
C16H36IN
140775
Tetrabutylammonium iodide, reagent grade, 98%
C16H36IN
359173
Tetrabutylammonium nitrate, 97%
C16H36N2O3
86893
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
C16H36ClNO4
86885
Tetrabutylammonium perchlorate, ≥98.0% (T)
C16H36ClNO4
86833
Tetrabutylammonium phosphate monobasic, puriss., ≥99.0% (T)
C16H38NO4P
268097
Tetrabutylammonium phosphate monobasic, 97%
C16H38NO4P
268100
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
C16H38NO4P