EMAIL THIS PAGE TO A FRIEND
Experimental lung research
Potential of sivelestat in protection against severe acute pancreatitis-associated lung injury in rats.
PMID 23005337
Abstract
The incidence of acute pancreatitis has been rising worldwide in the past few decades. Despite extensive research efforts, the population-based mortality from acute pancreatitis remains high. Since dysfunction of multiple vital organs, most importantly the lungs, is the major cause of early death in acute pancreatitis patients, developing effective strategies to manage lung injury has become one of the focuses of recent research efforts aiming at improving the outcome of patients with acute pancreatitis. In this study, we attempted to create a rat model of acute pancreatitis through intraductal infusion of taurocholate and to evaluate the potential of sivelestat, a synthetic neutrophil elastase inhibitor, in protection against acute pancreatitis-associated lung injury using this rat model. The results demonstrated that: (1) 5% sodium taurocholate successfully induced histopathologic and biochemical abnormalities in the circulation, lung and pancreas characteristic of human acute pancreatitis, including an increase in amylase concentration and a decrease in partial arterial oxygen pressure (PaO2) in the blood, increases in activities of myeloperoxidase (MPO) (a lung injury marker) and neutrophil elastase (a quantitative indicator of neutrophil infiltration), and levels of malondialdehyde (an indicator of lipid peroxidation) and tumor necrosis factor-alpha (a major inflammatory mediator) in the lung; (2) intravenous administration of sivelestat effectively attenuated the taurocholate-induced abnormalities in all parameters analyzed except for serum amylase concentration. Our findings have validated the taurocholate model of acute pancreatitis and demonstrated great therapeutic potential for sivelestat in managing acute pancreatitis-associated lung injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7198
Sivelestat sodium salt hydrate, ≥98% (HPLC), solid
C20H22N2O7S · xNa+ · yH2O