EMAIL THIS PAGE TO A FRIEND
Journal of pediatric hematology/oncology
Life-threatening agranulocytosis, anemia, and plasmacytosis after dipyrone use for fever in a child.
PMID 23007418
Abstract
Dipyrone or metamizole Na (Novalgin) is commonly used as an antipyretic, analgesic, and spasmolytic agent in some parts of the world; however, it is banned in developed countries because of severe side effects. Here we present a case of a 4-year-old boy who developed life-threatening agranulocytosis, anemia, and marked plasmacytosis in his bone marrow after dipyrone use for fever, which resolved with steroid therapy.