EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Facile synthesis of cysteine and triethanolamine capped CdTe nanoparticles.
PMID 23010054
Abstract
Cysteine and triethanolamine capped CdTe nanoparticles have been synthesized using a simple aqueous solution based method. This method involves the reaction of tellurium powder with sodium borohydride (NaBH(4)) in water to produce telluride ions (Te(2-)), followed by the simultaneous addition of an aqueous solution of cadmium chloride or other cadmium source (acetate, carbonate and nitrate) and solution of L-cysteine ethyl ester hydrochloride or triethanolamine. The effect of capping agent on the size, structure and morphology of the as-synthesized nanoparticles was investigated. The particles were characterized using optical spectroscopy, transmission electron microscopy (TEM), high-resolution TEM (HRTEM), X-ray diffraction (XRD) and Fourier transform infrared (FT-IR) spectroscopy.
Related Materials