EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Frequency-invariant temporal ordering of interneuronal discharges during hippocampal oscillations in awake mice.
PMID 23010933