EMAIL THIS PAGE TO A FRIEND
Cardiovascular drugs and therapy
Torsades de Pointes in the Guinea-pig heart : editorial to: "dofetilide promotes repolarization abnormalities in perfused Guinea-pig heart" by O.E. Osadchii.
PMID 23011586