EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
N-(Diazoacetyl)oxazolidin-2-thiones as sulfur-donor reagents: asymmetric synthesis of thiiranes from aldehydes.
PMID 23011905
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
128252
Ethylene sulfide, 98%
C2H4S