EMAIL THIS PAGE TO A FRIEND
Endoscopy
Tissue hyperplasia following placement of a biodegradable stent for a refractory esophageal stricture: treatment with argon plasma coagulation.
PMID 23012019
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
719846
Resomer® X 206 S, poly(dioxanone)