EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Draft genome sequence of Halomonas smyrnensis AAD6T.
PMID 23012275
Abstract
Halomonas smyrnensis AAD6(T) is a Gram-negative, aerobic, exopolysaccharide-producing, and moderately halophilic bacterium that produces levan, a fructose homopolymer with many potential uses in various industries. We report the draft genome sequence of H. smyrnensis AAD6(T), which will accelerate research on the rational design and optimization of microbial levan production.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L8647 Levan from Erwinia herbicola