EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Genome sequence of a cold-adaptable sulfamethoxazole-degrading bacterium, Pseudomonas psychrophila HA-4.
PMID 23012293
Abstract
Pseudomonas psychrophila HA-4 is a cold-adaptable, sulfamethoxazole-degrading bacterium. The genes related to its cold adaptation mechanism and sulfamethoxazole metabolism were unknown. We present the draft genome of strain HA-4. It could provide further insight into the sulfamethoxazole-degrading mechanism of strain HA-4.