EMAIL THIS PAGE TO A FRIEND
Water environment research : a research publication of the Water Environment Federation
An alternative process to treat boiler feed water for reuse.
PMID 23012772
Abstract
A bench-scale process to treat boiler feed water for reuse in steam generation was developed. Industrial water samples from a steam-assisted gravity drainage plant in northern Alberta, Canada, were obtained and samples characterized. The technology, which consists of coagulation-settling to remove oil/grease and particulates followed by an advanced oxidative treatment, led to clean water samples with negligible organic carbon. Coagulation followed by settling removed most particulates and some insoluble organics. The advanced oxidative treatment removed any remaining color in the samples, decreased the organic content to near-zero, and provided water ready for reuse.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
70340 Naphthenic acid, technical