EMAIL THIS PAGE TO A FRIEND
American journal of veterinary research
Effects of sterilization with hydrogen peroxide gas plasma, ethylene oxide, and steam on bioadhesive properties of nylon and polyethylene lines used for stabilization of canine stifle joints.
PMID 23013195
Abstract
To compare effects of sterilization with hydrogen peroxide gas plasma (HPGP), ethylene oxide, and steam on bioadhesive properties of nylon and polyethylene lines used for stabilization of canine stifle joints. Samples of a 36.3-kg test nylon leader line, 57.8-kg test nylon fishing line, and 2-mm ultrahigh-molecular weight polyethylene (UHMWPE) were used. In this in vitro study, samples of nylon leader line, fishing line, and UHMWPE sterilized by use of HPGP, ethylene oxide, and steam or unsterilized samples were used. Bacterial adherence on unsterilized and sterilized samples was tested with Staphylococcus epidermidis and Escherichia coli. Five samples were examined for each line type and sterilization condition, and final colony counts were obtained. Bacterial adherence was significantly affected by method of sterilization for all 3 line types. For most of the samples, bacterial adherence was similar or lower when HPGP sterilization was used, compared with results for sterilization via ethylene oxide and steam, respectively. Bacterial adherence was significantly higher for UHMWPE, compared with adherence for the nylon line, regardless of the sterilization method used. Bacterial adherence was higher for nylon fishing line than for nylon leader line for S epidermidis after ethylene oxide sterilization and for E coli after HPGP and ethylene oxide sterilization. Effects of HPGP sterilization on bioadhesive properties of nylon and polyethylene lines compared favorably with those for ethylene oxide and steam sterilization. Also, nylon line may be a more suitable material than UHMWPE for suture prostheses on the basis of bacterial adherence properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48838
Ethylene oxide solution, certified reference material, 50 mg/mL in methanol
C2H4O