EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
[5] Determination of cystine and cysteine as S-sulfocysteine.
PMID 23014392
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72030
Sodium tetrathionate dihydrate, purum p.a., ≥98.0% (T)
Na2O6S4 · 2H2O