EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Mesoporous nanomagnet supercaptors for selective heme-proteins from human cells.
PMID 23014515
Abstract
Nanomagnet-selective supercaptors of heme-proteins (iron-porphyrin prosthetic group) based on mesoporous NiO and Fe(3)O(4) NPs were fabricated.