EMAIL THIS PAGE TO A FRIEND
AIDS (London, England)
No impact of rifamycin selection on tuberculosis treatment outcome in HIV coinfected patients.
PMID 23014518
Abstract
Rifabutin has been substituted for rifampicin when treating tuberculosis (TB)/HIV coinfection. However, despite reports of anti-TB treatment failure and acquired rifamycin resistance, long-term clinical outcome data are lacking. Observational analyses performed in a UK TB/HIV cohort demonstrated no difference in severe adverse events, anti-TB treatment completion, relapse frequency or subsequent rifamycin resistance when rifampicin and rifabutin were compared, using different combinations of antiretroviral therapy. Our data support the wider use of rifabutin in TB/HIV coinfection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R3530 Rifabutin, >98% (HPLC), powder
C46H62N4O11
R8626
Rifamycin SV sodium salt, potency: ≥900 units (dry basis)
C37H46NNaO12