EMAIL THIS PAGE TO A FRIEND
Heart advisor
I am a 37-year-old woman and was diagnosed with coronary artery spasms two years ago. I am taking Plavix, verapamil and Imdur for my condition. What other things can I do to help my heart?
PMID 23016179