EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Cytotoxic platinum(II) intercalators that incorporate 1R,2R-diaminocyclopentane.
PMID 23018340
Abstract
Twelve metallointercalators of the type [Pt(I(L))(A(L))](2+), where A(L) is either the R,R or S,S enantiomer of 1,2-diaminocyclopentane (DACP) and I(L) is either 1,10-phenathroline, 4-methyl-1,10-phenanthroline, 5-methyl-1,10-phenanthroline, 4,7-dimethyl-1,10-phenanthroline, 5,6-dimethyl-1,10-phenanthroline or 3,4,7,8-tetramethyl-1,10-phenanthroline, were synthesised, characterised and the cytotoxicity to the L1210 cell line was determined. The crystal structures of PHENRRDACP and PHENSS were obtained as monoclinic with a space group of P2(1) (a/Å = 11.4966, b/Å = 6.6983, c/Å = 12.0235) and P2(1) (a/Å = 11.5777, b/Å = 7.0009, c/Å = 12.5079), respectively. The R,R enantiomer of 1,2-diaminocyclopentane (RRDACP) produced the most cytotoxic metallointercalators. The most cytotoxic metallointercalators were 56MERRDACP and 47MERRDACP with IC(50) values of 0.16 and 0.17 μM, respectively, in comparison to cisplatin (1 μM).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
230847
5-Methyl-1,10-phenanthroline, ≥99%
C13H10N2