EMAIL THIS PAGE TO A FRIEND
Journal of pediatric hematology/oncology
Hemorrhagic shock due to spontaneous rupture of adrenal neuroblastoma in an infant: a rare case and review of the literature.
PMID 23018564
Abstract
Spontaneous rupture of adrenal neuroblastoma is very rare in infants, in contrast to neonates. This report describes a 9-month-old boy presenting with acute hemorrhagic shock due to spontaneous rupture of adrenal neuroblastoma. MYCN oncogene amplification may be a predisposing factor for spontaneous rupture and bleeding of neuroblastoma. An appropriate surgical treatment for this condition must be discussed according to the patient's general state and the tumor features, such as staging, the origin, and local invasiveness.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6069
Thio-TEPA, 98%, solid
C6H12N3PS