EMAIL THIS PAGE TO A FRIEND
Archives of disease in childhood
Effect of artificial food colours on childhood behaviour.
PMID 2301986
Abstract
We performed an objective evaluation of 39 children whose behaviour was observed by their parents to improve on an artificial food additive free diet and to deteriorate with dietary lapses. Only 19 children completed a double blind placebo controlled challenge study with artificial food colours. In these children food colours were shown to have an adverse effect on a daily Conners' rating of behaviour, but most parents could not detect these changes. A pharmacological mechanism of food additive intolerance is proposed to explain these effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52245
Carmoisine, analytical standard
C20H12N2Na2O7S2
214515
Chromotrope FB, Dye content 50 %
C20H12N2Na2O7S2