EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Variation of icariin and total flavonoid of Epimedium acuminatum in different parts and habitats].
PMID 23019870
Abstract
Through comparative study on contents of icariin and total flavonoid of Epimedium acuminatum in different habitats and parts, the distribution and correlation of the two components were observed. Twenty-four sample spots in four habitats were set up, and the whole plant of these samples was divided as following: leaf, root, stem and rhizome. Total flavonoid and icariin of samples were determined by using the UV method and the HPLC, respectively. The data was analyzed by SPSS 17.0. The distribution of icariin in different parts had the pattern: leaf > root > stem > rhizome, the total flavonoid content was higher in leaf, but it showed no difference in other parts. Habitat had certain effect on icariin accumulation in E. acuminatum. The significant differences of total flavonoid content in E. acuminatum from different parts were not observed. The distribution of icariin from different parts and habitats has high selectivity. Metabolism and accumulation of flavonoid content in each part have no difference. Flavonoid content is less affected by environment. Considering the distribution of icariin and flavonoid content in every part and the growth strategy in different habitats comprehensively, it is reasonable to harvest the aerial part for the sustainable utilization of E. acuminatum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I1286
Icariin, ≥94% (HPLC)
C33H40O15