EMAIL THIS PAGE TO A FRIEND
Langmuir : the ACS journal of surfaces and colloids
Enantiospecific desorption of R- and S-propylene oxide from D- or L-lysine modified Cu(100) surfaces.
PMID 23020648
Abstract
The enantiospecific desorption kinetics of R- and S-propylene oxide (PO) from a Cu(100) surface modified by enantiomerically pure D- or L-lysine have been studied using temperature programmed desorption. These experiments have used R- or S-PO as the chiral probe for study of enantiospecific adsorption on Cu(100) surfaces modified with D- or L-lysine. This chiral probe/modifier/Cu system manifests a significant diastereomeric effect in the R- and S-PO peak desorption temperatures and, hence, true enantiospecific behavior. The enantiospecificity in the PO desorption kinetics is observed only over a narrow range of lysine modifier coverage with a maximum at a lysine coverage leaving an empty site density of θ(O) ≈ 0.25. The observation of enantiospecific behavior in the PO/lysine/Cu(100) system is in contrast with the failed results of prior attempts to observe enantiospecific desorption from chirally modified Cu surfaces. The potential for hydrogen-bonding interactions between the chiral probe and chiral modifier, which can depend on the coverage and configuration of the adsorbed modifier, may play a crucial role in enantiospecific adsorption on lysine modified Cu surfaces.