EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Cyclic tetrapeptides with thioacetate tails or intramolecular disulfide bridge as potent inhibitors of histone deacetylases.
PMID 23021104
Abstract
Two thioacetate tails were introduced to the chlamydocin- and CHAP31-related cyclic tetrapeptides. An intramolecular disulfide bridge could be formed in the CHAP31-related cyclic peptides. Both the thioacetate-tailed and disulfide-bridged peptides were potent histone deacetylase inhibitors in the presence of sulfhydryl compound. Potent p21 promoter inducing activity was also observed in vivo.
Related Materials