EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
L-type Ca(2+) channels, Ca(2+)-induced Ca(2+) release, and BK(Ca) channels in airway stretch-induced contraction.
PMID 23022333
Abstract
Resistance arteries constrict in response to mechanical stress. This response is myogenic in nature, and reliant on membrane depolarization, activation of L-type Ca(2+) channels, Ca(2+)-induced Ca(2+)-release and large conductance Ca(2+)-dependent K(+) channels (BK(Ca)). Airway smooth muscle is also affected by mechanical stress: a deep inspiration produces a bronchodilation in healthy individuals, but bronchoconstriction in moderate to severe asthmatics. In this study, our objective was to investigate the regulation of this airway stretch-activated contractile response (R(stretch)), and explore its similarities to the vascular myogenic response. Using a pharmacological approach in intact bovine bronchial segments cannulated horizontally in an organ bath, we showed the ability of carbachol (2-carbamoyloxyethyl-trimethyl-azanium), KCl, neurokinin-A, and U46619 (9,11-dideoxy-9α,11α-methanoepoxy-prosta-5Z, 13E-dien-1-oic acid) to generate R(stretch) in a concentration-dependent manner. R(stretch) was significantly reduced by nifedipine, ryanodine, and iberiotoxin, suggesting that it possesses characteristics similar to those of the vascular smooth muscle myogenic response, such as a role for membrane depolarization, L-type Ca(2+) channel, ryanodine receptors and BK(Ca) channel activation. This study demonstrates a novel role for the L-type Ca(2+) channel in airway smooth muscle and provides new insights into possible mechanisms regulating the deep inspiration-induced bronchoconstriction seen in asthmatics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B133
(S)-(−)-Bay K8644, ≥98% (HPLC), solid
C16H15F3N2O4