EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Structure, DNA binding and cleavage of a new Zn(II)Mn(II) macrocyclic complex.
PMID 23022611
Abstract
A new heterodinuclear complex of an unsymmetrical macrocycle [ZnMnL(CH(3)O)(2)]·H(2)O has been synthesized by the cyclocondensation between N,N'-bis(3-formyl-5-chlorosalicylidene)ethylenediimine and 2-hydroxyl-1,3-propanediamine in the presence of the metal ions, and characterized by elemental analyses, IR spectra and X-ray determination. The interactions of the complex with DNA have been investigated by UV absorption, fluorescence spectroscopy, viscosity measurements and electrochemical studies. Absorption spectroscopic investigation reveals that the complex has good binding propensity to calf thymus DNA by intercalation with a binding constant of 2.52 × 10(5)M(-1). Fluorescence spectroscopy shows that the complex can displace ethidium bromide and bind to DNA, with a quenching constant of 4.37 × 10(3)M(-1). The agarose gel electrophoresis studies show that pBR322 plasmid DNA can be transformed to nicked form and linear form in air by the complex.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D23807
1,3-Diaminopropane dihydrochloride, 98%
C3H10N2 · 2HCl