EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis, antitumor, cytotoxic and antioxidant evaluation of some new pyrazolotriazines attached to antipyrine moiety.
PMID 23022735
Abstract
Iminopropanehydrazonoyl cyanide 4 was achieved upon reaction of antipyrine diazonium salt 2 with 3-iminobutanenitrile (3) in EtOH/AcONa. 3-Aminopyrazole derivative 5 was obtained upon reaction of 4 with hydrazine hydrate. Diazodization of 5 afforded the diazonium salt 6 which coupled with active methylene compounds 7-10, 19, 20, 25, 29 and 32 in pyridine to give aryl hydrazone derivatives 11-14, 21, 22, 26, 30 and 33, respectively. Refluxing of compounds 11-14, 21, 22, 26 and 33 in acetic acid afforded the pyrazolotriazines 15-18, 23, 24, 28 and 35, respectively. The newly synthesized compounds were screened for their cytotoxic and antioxidant activities. The results showed clearly that compounds 4, 5, 13, 22, and 24 displayed promising in vitro anticancer activity against four different cell lines (HepG2, WI 38, VERO and MCF-7). Compounds 4 and 22 are the more potent antioxidant and anticancer agents. On the other hand, most of the compounds exhibited good cytotoxic activity toward (EAC).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10784
Antipyrine, tested according to Ph.Eur.
C11H12N2O