EMAIL THIS PAGE TO A FRIEND
The international journal of occupational and environmental medicine
Occupational exposure of gasoline station workers to BTEX compounds in Bangkok, Thailand.
PMID 23022861
Abstract
Gasoline station workers are exposed to volatile organic compounds such as benzene, toluene, ethylbenzene and xylene (BTEX). To determine the level of exposure to BTEX compounds among gasoline station workers and measure the roadside concentrations of these compounds in the inner and outer areas of Bangkok, Thailand. 49 workers at 6 gasoline stations in the inner and outer areas of Bangkok participated in this study. Samples of ambient air were collected from the area near gas pumps at each station and at the roadside in front of the gas stations by charcoal tubes. All samples were analyzed for BTEX compounds by gas chromatography-flame ionized detector (GC-FID). The mean BTEX concentration in gas stations was slightly higher than that of the roadside; there was no significant difference in the concentration between inner and outer areas. The mean lifetime cancer risks for workers exposed to benzene and ethylbenzene for 30 years were estimated at 1.75×10⁻⁴ and 9.55×10⁻⁷. The estimated hazard quotients for BTEX compounds were 0.600, 0.008, 0.007 and 0.002, respectively. The most prevalent symptoms of workers were headache (61%), fatigue (29%) and throat irritation (11%), respectively. Exposure to benzene and toluene was significantly associated with fatigue (p<0.05). Exposure to BTEX compounds would increase the risk of cancer in gasoline station workers. Exposure to benzene and toluene may cause fatigue.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03079
Ethylbenzene, analytical standard
C8H10
296848
Ethylbenzene, anhydrous, 99.8%
C8H10
E12508
Ethylbenzene, ReagentPlus®, 99%
C8H10
40036
Ethylbenzene solution, certified reference material, 5000 μg/mL in methanol
C8H10
612936
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 3 mm × 8 in.
C8H10
551341
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
C8H10
487112
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 3 mm × 8 in.
C8H10
487104
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
C8H10
487120
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.
C8H10
612065
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.
C8H10
612480
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, chromium(III) acetylacetonate 0.1 mg/mL, NMR tube size 5 mm × 8 in.
C8H10
708046
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D)
C8H10
708054
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d ("100%", 99.96 atom % D)
C8H10
710725
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.
C8H10
714917
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
C8H10
717967
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
C8H10
717959
Ethylbenzene solution, NMR reference standard, 1% in chloroform-d (99.8 atom % D)
C8H10
736198
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
C8H10
733768
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
C8H10
721018
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.
C8H10
721026
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
C8H10
720291
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
C8H10