EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Metal-free direct amination/aromatization of 2-cyclohexenones to iodo-N-arylanilines and N-arylanilines promoted by iodine.
PMID 23023304
Abstract
An iodine mediated aromatization leading to a one-pot synthesis of iodo-N-arylanilines and N-arylanilines is reported. This highly regioselective aliphatic-aromatic transformation can be performed with various combinations of 2-cyclohexenones and anilines. The presence of a directing group is crucial for achieving high yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C102814
2-Cyclohexen-1-one, ≥95%
C6H8O