EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
In situ formation of β-glycosyl imidinium triflate from participating thioglycosyl donors: elaboration to disarmed-armed iterative glycosylation.
PMID 23023321
Abstract
β-Glycosyl imidinium triflate is generated from participating thioglycoside donors for disarmed-armed iterative glycosylations and one-pot oligosaccharide synthesis.