EMAIL THIS PAGE TO A FRIEND
Deutsche medizinische Wochenschrift (1946)
[Irritable bowel disease: recent developments].
PMID 23023617
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33999
Rifaximin, VETRANAL, analytical standard
C43H51N3O11