EMAIL THIS PAGE TO A FRIEND
Journal of Zhejiang University. Science. B
Extraction and isolation of the salidroside-type metabolite from zinc (Zn) and cadmium (Cd) hyperaccumulator Sedum alfredii Hance.
PMID 23024051
Abstract
The active metabolite in the post-harvested biomass of zinc (Zn) and cadmium (Cd) hyperaccumulator Sedum alfredii Hance from phytoextraction is of great interest in China. The current study demonstrates that a salidroside-type metabolite can be yielded from the Zn/Cd hyperaccumulator S. alfredii biomass by means of sonication/ethanol extraction and macroporous resin column (AB-8 type) isolation. The concentrations of Zn and Cd in the salidroside-type metabolite were below the limitation of the national standards.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43866
Salidroside, analytical standard
C14H20O7